Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607754 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKRN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MKRN1 - makorin ring finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145125.1 | Intron | NP_001138597.1 | ||||
NM_001291663.1 | Intron | NP_001278592.1 | ||||
NM_013446.3 | Intron | NP_038474.2 | ||||
XM_011515996.1 | Intron | XP_011514298.1 | ||||
XM_011515997.2 | Intron | XP_011514299.1 | ||||
XM_011515998.1 | Intron | XP_011514300.1 |
Set Membership: |
HapMap Validated |