Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610089 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFCAB10 - EF-hand calcium binding domain 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RINT1 - RAD50 interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021930.4 | Intron | NP_068749.3 | ||||
XM_005250524.3 | Intron | XP_005250581.1 | ||||
XM_011516458.2 | Intron | XP_011514760.1 | ||||
XM_017012496.1 | Intron | XP_016867985.1 | ||||
XM_017012497.1 | Intron | XP_016867986.1 | ||||
XM_017012498.1 | Intron | XP_016867987.1 |