Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611838 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MITD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MITD1 - microtubule interacting and trafficking domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320417.1 | 57 | Intron | NP_001307346.1 | |||
NM_001320418.1 | 57 | Intron | NP_001307347.1 | |||
NM_001320419.1 | 57 | Intron | NP_001307348.1 | |||
NM_138798.2 | 57 | Intron | NP_620153.1 | |||
XM_011510581.2 | 57 | Intron | XP_011508883.1 | |||
XM_011510582.2 | 57 | Intron | XP_011508884.1 | |||
XM_011510583.1 | 57 | Intron | XP_011508885.1 | |||
XM_017003314.1 | 57 | Intron | XP_016858803.1 | |||
XM_017003315.1 | 57 | Intron | XP_016858804.1 |
MRPL30 - mitochondrial ribosomal protein L30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145212.3 | 57 | UTR 5 | NP_660213.1 |
Set Membership: |
HapMap Validated |