Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147561 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGB5 - integrin subunit beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002213.4 | Intron | NP_002204.2 | ||||
XM_005247436.2 | Intron | XP_005247493.1 | ||||
XM_006713630.2 | Intron | XP_006713693.1 | ||||
XM_017006351.1 | Intron | XP_016861840.1 | ||||
XM_017006352.1 | Intron | XP_016861841.1 | ||||
XM_017006353.1 | Intron | XP_016861842.1 | ||||
XM_017006354.1 | Intron | XP_016861843.1 | ||||
XM_017006355.1 | Intron | XP_016861844.1 |
Set Membership: |
HapMap |