Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605984 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EED PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EED - embryonic ectoderm development | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308007.1 | 563 | UTR 5 | NP_001294936.1 | |||
NM_003797.4 | 563 | UTR 5 | NP_003788.2 | |||
XM_005274373.2 | 563 | UTR 5 | XP_005274430.1 | |||
XM_011545330.1 | 563 | UTR 5 | XP_011543632.1 | |||
XM_011545331.1 | 563 | UTR 5 | XP_011543633.1 | |||
XM_017018512.1 | 563 | UTR 5 | XP_016874001.1 | |||
XM_017018513.1 | 563 | UTR 5 | XP_016874002.1 |
Set Membership: |
HapMap |