Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615038 MIM: 603996 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC114 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC114 - coiled-coil domain containing 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144577.3 | 2654 | UTR 3 | NP_653178.3 | |||
XM_005259413.3 | 2654 | Intron | XP_005259470.1 | |||
XM_005259414.3 | 2654 | Intron | XP_005259471.1 | |||
XM_005259415.3 | 2654 | Intron | XP_005259472.1 | |||
XM_005259416.4 | 2654 | Intron | XP_005259473.1 | |||
XM_011527515.2 | 2654 | UTR 3 | XP_011525817.1 | |||
XM_011527516.2 | 2654 | UTR 3 | XP_011525818.1 | |||
XM_011527517.1 | 2654 | Intron | XP_011525819.1 | |||
XM_017027483.1 | 2654 | Intron | XP_016882972.1 |
ZNF114 - zinc finger protein 114 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |