Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602965 MIM: 610728 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FABP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FABP7 - fatty acid binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319039.1 | 243 | Missense Mutation | ACG,ATG | T,M 61 | NP_001305968.1 | |
NM_001319041.1 | 243 | Missense Mutation | ACG,ATG | T,M 61 | NP_001305970.1 | |
NM_001319042.1 | 243 | Missense Mutation | ACG,ATG | T,M 57 | NP_001305971.1 | |
NM_001446.4 | 243 | Missense Mutation | ACG,ATG | T,M 61 | NP_001437.1 |
SMPDL3A - sphingomyelin phosphodiesterase acid like 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |