Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608714 | ||||||||||||||||||||
Literature Links: |
SNTG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNTG1 - syntrophin gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287813.2 | Intron | NP_001274742.1 | ||||
NM_001287814.2 | Intron | NP_001274743.1 | ||||
NM_001321773.1 | Intron | NP_001308702.1 | ||||
NM_001321775.1 | Intron | NP_001308704.1 | ||||
NM_001321776.1 | Intron | NP_001308705.1 | ||||
NM_001321777.1 | Intron | NP_001308706.1 | ||||
NM_001321778.1 | Intron | NP_001308707.1 | ||||
NM_018967.4 | Intron | NP_061840.1 | ||||
XM_011517548.2 | Intron | XP_011515850.1 | ||||
XM_017013579.1 | Intron | XP_016869068.1 | ||||
XM_017013580.1 | Intron | XP_016869069.1 | ||||
XM_017013581.1 | Intron | XP_016869070.1 | ||||
XM_017013582.1 | Intron | XP_016869071.1 |