Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602846 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UBE2K - ubiquitin conjugating enzyme E2 K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111112.1 | Intron | NP_001104582.1 | ||||
NM_001111113.1 | Intron | NP_001104583.1 | ||||
NM_001312646.1 | Intron | NP_001299575.1 | ||||
NM_001312647.1 | Intron | NP_001299576.1 | ||||
NM_001312648.1 | Intron | NP_001299577.1 | ||||
NM_005339.4 | Intron | NP_005330.1 |
Set Membership: |
HapMap Validated |