Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGGAGGGGCCTGTCCGCCCTC[G/T]TTCCCCCCAGAGCTACAGGACTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604176 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928674 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101928674 - uncharacterized LOC101928674 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOCS3 - suppressor of cytokine signaling 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003955.4 | 1215 | UTR 3 | NP_003946.3 |
Set Membership: |
HapMap Validated |