Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606740 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF180 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF180 - zinc finger protein 180 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278508.2 | 2645 | UTR 3 | NP_001265437.2 | |||
NM_001278509.2 | 2645 | UTR 3 | NP_001265438.2 | |||
NM_001288759.2 | 2645 | UTR 3 | NP_001275688.2 | |||
NM_001288760.2 | 2645 | UTR 3 | NP_001275689.1 | |||
NM_001288761.2 | 2645 | UTR 3 | NP_001275690.1 | |||
NM_001288762.2 | 2645 | UTR 3 | NP_001275691.1 | |||
NM_001291633.1 | 2645 | UTR 3 | NP_001278562.1 | |||
NM_013256.5 | 2645 | UTR 3 | NP_037388.3 | |||
XM_011527280.2 | 2645 | UTR 3 | XP_011525582.1 | |||
XM_017027250.1 | 2645 | UTR 3 | XP_016882739.1 | |||
XM_017027251.1 | 2645 | UTR 3 | XP_016882740.1 | |||
XM_017027252.1 | 2645 | UTR 3 | XP_016882741.1 | |||
XM_017027253.1 | 2645 | UTR 3 | XP_016882742.1 | |||
XM_017027254.1 | 2645 | UTR 3 | XP_016882743.1 | |||
XM_017027255.1 | 2645 | UTR 3 | XP_016882744.1 | |||
XM_017027256.1 | 2645 | UTR 3 | XP_016882745.1 |
Set Membership: |
HapMap |