Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602113 MIM: 602742 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
KMT2D - lysine methyltransferase 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003482.3 | 44 | Intron | NP_003473.3 | |||
XM_005269162.4 | 44 | Intron | XP_005269219.1 | |||
XM_006719614.3 | 44 | Intron | XP_006719677.1 | |||
XM_006719616.3 | 44 | Intron | XP_006719679.1 | |||
XM_011538770.2 | 44 | Intron | XP_011537072.1 | |||
XM_011538771.2 | 44 | Intron | XP_011537073.1 | |||
XM_011538772.2 | 44 | Intron | XP_011537074.1 | |||
XM_011538773.2 | 44 | Intron | XP_011537075.1 | |||
XM_011538774.2 | 44 | Intron | XP_011537076.1 | |||
XM_011538776.2 | 44 | Intron | XP_011537078.1 |
PRKAG1 - protein kinase AMP-activated non-catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206709.1 | 44 | Intron | NP_001193638.1 | |||
NM_001206710.1 | 44 | Intron | NP_001193639.1 | |||
NM_002733.4 | 44 | Intron | NP_002724.1 | |||
XM_005269019.4 | 44 | Silent Mutation | XP_005269076.1 | |||
XM_005269020.2 | 44 | Intron | XP_005269077.1 | |||
XM_006719499.2 | 44 | Intron | XP_006719562.1 | |||
XM_011538562.2 | 44 | Intron | XP_011536864.1 | |||
XM_017019622.1 | 44 | Intron | XP_016875111.1 | |||
XM_017019623.1 | 44 | Intron | XP_016875112.1 |
Set Membership: |
HapMap |