Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604068 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPD52 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TPD52 - tumor protein D52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025252.2 | Intron | NP_001020423.1 | ||||
NM_001025253.2 | Intron | NP_001020424.1 | ||||
NM_001287140.1 | Intron | NP_001274069.1 | ||||
NM_001287142.1 | Intron | NP_001274071.1 | ||||
NM_001287143.1 | Intron | NP_001274072.1 | ||||
NM_001287144.1 | Intron | NP_001274073.1 | ||||
NM_005079.3 | Intron | NP_005070.1 |