Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTATACAGACATAATAAATCTG[A/G]TTGTTTCATAGCGCTAACATAGAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612246 MIM: 604524 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD302 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CD302 - CD302 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY75 - lymphocyte antigen 75 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002349.3 | Intron | NP_002340.2 |
LY75-CD302 - LY75-CD302 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198759.1 | Intron | NP_001185688.1 | ||||
NM_001198760.1 | Intron | NP_001185689.1 |