Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613446 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP120 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CEP120 - centrosomal protein 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166226.1 | 3256 | Missense Mutation | ATA,GTA | I,V 910 | NP_001159698.1 | |
NM_153223.3 | 3256 | Missense Mutation | ATA,GTA | I,V 936 | NP_694955.2 | |
XM_005271901.4 | 3256 | Missense Mutation | ATA,GTA | I,V 891 | XP_005271958.1 | |
XM_011543185.2 | 3256 | Missense Mutation | ATA,GTA | I,V 910 | XP_011541487.1 | |
XM_011543186.2 | 3256 | Missense Mutation | ATA,GTA | I,V 447 | XP_011541488.1 | |
XM_017009085.1 | 3256 | Missense Mutation | ATA,GTA | I,V 447 | XP_016864574.1 |