Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAAATAATTGCATGAGTAAAT[A/G]ATTTCAAAACTGAGATAAATGTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612014 | ||||||||||||||||||||
Literature Links: |
LOC100506124 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100506124 - uncharacterized LOC100506124 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC21B - tetratricopeptide repeat domain 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024753.4 | 7148 | Intron | NP_079029.3 | |||
XM_006712761.1 | 7148 | Intron | XP_006712824.1 | |||
XM_011511871.2 | 7148 | Intron | XP_011510173.1 | |||
XM_011511872.2 | 7148 | Intron | XP_011510174.1 | |||
XM_017004967.1 | 7148 | UTR 3 | XP_016860456.1 | |||
XM_017004968.1 | 7148 | Intron | XP_016860457.1 | |||
XM_017004969.1 | 7148 | Intron | XP_016860458.1 |