Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGAGCCGGGCGGCGCAGGCGCAG[A/T]GTCCCCGGGCCAAGATGGCTGCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611668 MIM: 608382 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CORO7 - coronin 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CORO7-PAM16 - CORO7-PAM16 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAJA3 - DnaJ heat shock protein family (Hsp40) member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135110.2 | 63 | UTR 5 | NP_001128582.1 | |||
NM_001286516.1 | 63 | UTR 5 | NP_001273445.1 | |||
NM_005147.5 | 63 | UTR 5 | NP_005138.3 |
Set Membership: |
HapMap |