Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607533 MIM: 607532 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIS3 - DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIBF1 - progesterone immunomodulatory binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006346.2 | Intron | NP_006337.2 | ||||
XM_011534881.2 | Intron | XP_011533183.1 | ||||
XM_011534882.2 | Intron | XP_011533184.1 | ||||
XM_011534884.2 | Intron | XP_011533186.1 | ||||
XM_011534885.2 | Intron | XP_011533187.1 | ||||
XM_011534886.2 | Intron | XP_011533188.1 | ||||
XM_017020350.1 | Intron | XP_016875839.1 | ||||
XM_017020351.1 | Intron | XP_016875840.1 | ||||
XM_017020352.1 | Intron | XP_016875841.1 | ||||
XM_017020353.1 | Intron | XP_016875842.1 | ||||
XM_017020354.1 | Intron | XP_016875843.1 |