Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612189 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PBLD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PBLD - phenazine biosynthesis like protein domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033083.1 | 1125 | UTR 3 | NP_001028255.1 | |||
NM_022129.3 | 1125 | Intron | NP_071412.2 | |||
XM_005270028.3 | 1125 | Intron | XP_005270085.1 | |||
XM_011540060.2 | 1125 | Intron | XP_011538362.1 | |||
XM_017016513.1 | 1125 | Intron | XP_016872002.1 | |||
XM_017016514.1 | 1125 | Intron | XP_016872003.1 |