Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300269 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC8 - histone deacetylase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166418.1 | Intron | NP_001159890.1 | ||||
NM_001166419.1 | Intron | NP_001159891.1 | ||||
NM_001166420.1 | Intron | NP_001159892.1 | ||||
NM_001166422.1 | Intron | NP_001159894.1 | ||||
NM_001166448.1 | Intron | NP_001159920.1 | ||||
NM_018486.2 | Intron | NP_060956.1 | ||||
XM_011530986.2 | Intron | XP_011529288.2 | ||||
XM_011530987.2 | Intron | XP_011529289.2 | ||||
XM_017029640.1 | Intron | XP_016885129.1 | ||||
XM_017029641.1 | Intron | XP_016885130.1 | ||||
XM_017029642.1 | Intron | XP_016885131.1 | ||||
XM_017029643.1 | Intron | XP_016885132.1 | ||||
XM_017029644.1 | Intron | XP_016885133.1 | ||||
XM_017029645.1 | Intron | XP_016885134.1 | ||||
XM_017029646.1 | Intron | XP_016885135.1 | ||||
XM_017029647.1 | Intron | XP_016885136.1 |