Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605610 MIM: 610195 | ||||||||||||||||||||
Literature Links: |
MIR4749 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4749 - microRNA 4749 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNKP - polynucleotide kinase 3'-phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTOV1 - prostate tumor overexpressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305105.1 | Intron | NP_001292034.1 | ||||
NM_001305108.1 | Intron | NP_001292037.1 | ||||
NM_017432.4 | Intron | NP_059128.2 | ||||
XM_005258998.4 | Intron | XP_005259055.1 | ||||
XM_006723244.3 | Intron | XP_006723307.1 | ||||
XM_011527033.1 | Intron | XP_011525335.1 | ||||
XM_011527034.2 | Intron | XP_011525336.1 | ||||
XM_011527035.1 | Intron | XP_011525337.1 | ||||
XM_017026879.1 | Intron | XP_016882368.1 | ||||
XM_017026880.1 | Intron | XP_016882369.1 |
PTOV1-AS1 - PTOV1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTOV1-AS2 - PTOV1 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |