Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCAGGCTGAGAGCCTGTGGAGAC[A/G]GACGGGGCTGGGGCTGGCGGCCGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613713 MIM: 176895 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724474 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102724474 - WAS/WASL-interacting protein family member 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCID2 - PCI domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROZ - protein Z, vitamin K dependent plasma glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256134.1 | Intron | NP_001243063.1 | ||||
NM_003891.2 | Intron | NP_003882.1 | ||||
XM_017020812.1 | Intron | XP_016876301.1 | ||||
XM_017020813.1 | Intron | XP_016876302.1 |
Set Membership: |
HapMap |