Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 178640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFTPB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFTPB - surfactant protein B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000542.3 | 1633 | UTR 3 | NP_000533.3 | |||
NM_198843.2 | 1633 | Intron | NP_942140.2 | |||
XM_005264487.2 | 1633 | UTR 3 | XP_005264544.1 | |||
XM_005264488.3 | 1633 | UTR 3 | XP_005264545.2 | |||
XM_005264490.4 | 1633 | Intron | XP_005264547.2 | |||
XM_017004702.1 | 1633 | Intron | XP_016860191.1 | |||
XM_017004703.1 | 1633 | Intron | XP_016860192.1 |