Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 178640 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFTPB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SFTPB - surfactant protein B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000542.3 | 1517 | UTR 3 | NP_000533.3 | |||
NM_198843.2 | 1517 | Intron | NP_942140.2 | |||
XM_005264487.2 | 1517 | UTR 3 | XP_005264544.1 | |||
XM_005264488.3 | 1517 | UTR 3 | XP_005264545.2 | |||
XM_005264490.4 | 1517 | Intron | XP_005264547.2 | |||
XM_017004702.1 | 1517 | Intron | XP_016860191.1 | |||
XM_017004703.1 | 1517 | Intron | XP_016860192.1 |
Set Membership: |
HapMap |