Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605435 | ||||||||||||||||||||
Literature Links: |
PRKD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRKD1 - protein kinase D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002742.2 | Intron | NP_002733.2 | ||||
XM_005267859.1 | Intron | XP_005267916.1 | ||||
XM_011536964.2 | Intron | XP_011535266.1 | ||||
XM_011536965.2 | Intron | XP_011535267.1 | ||||
XM_017021461.1 | Intron | XP_016876950.1 | ||||
XM_017021462.1 | Intron | XP_016876951.1 |