Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602315 | ||||||||||||||||||||
Literature Links: |
MAP2K3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP2K3 - mitogen-activated protein kinase kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316332.1 | Intron | NP_001303261.1 | ||||
NM_002756.4 | Intron | NP_002747.2 | ||||
NM_145109.2 | Intron | NP_659731.1 | ||||
XM_005256723.2 | Intron | XP_005256780.1 | ||||
XM_011523958.2 | Intron | XP_011522260.1 | ||||
XM_011523959.1 | Intron | XP_011522261.1 | ||||
XM_017024857.1 | Intron | XP_016880346.1 | ||||
XM_017024858.1 | Intron | XP_016880347.1 | ||||
XM_017024859.1 | Intron | XP_016880348.1 |