Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLMN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLMN - glomulin, FKBP associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319683.1 | Intron | NP_001306612.1 | ||||
NM_053274.2 | Intron | NP_444504.1 | ||||
XM_005270401.3 | Intron | XP_005270458.1 | ||||
XM_006710309.2 | Intron | XP_006710372.1 | ||||
XM_011540546.2 | Intron | XP_011538848.1 | ||||
XM_017000137.1 | Intron | XP_016855626.1 | ||||
XM_017000138.1 | Intron | XP_016855627.1 | ||||
XM_017000139.1 | Intron | XP_016855628.1 | ||||
XM_017000140.1 | Intron | XP_016855629.1 | ||||
XM_017000141.1 | Intron | XP_016855630.1 | ||||
XM_017000142.1 | Intron | XP_016855631.1 | ||||
XM_017000143.1 | Intron | XP_016855632.1 | ||||
XM_017000144.1 | Intron | XP_016855633.1 |