Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604350 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB3D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAB3D - RAB3D, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004283.3 | 4098 | UTR 3 | NP_004274.1 |
TSPAN16 - tetraspanin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282509.1 | 4098 | Intron | NP_001269438.1 | |||
NM_001282510.1 | 4098 | Intron | NP_001269439.1 | |||
NM_012466.3 | 4098 | Intron | NP_036598.1 | |||
XM_011527901.1 | 4098 | Intron | XP_011526203.1 | |||
XM_011527902.1 | 4098 | Intron | XP_011526204.1 | |||
XM_011527903.2 | 4098 | Intron | XP_011526205.1 |