Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601826 MIM: 610570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKD - diacylglycerol kinase delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP40 - ubiquitin specific peptidase 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018218.2 | 4215 | UTR 3 | NP_060688.1 | |||
XM_006712612.3 | 4215 | UTR 3 | XP_006712675.1 | |||
XM_011511396.2 | 4215 | UTR 3 | XP_011509698.1 | |||
XM_011511397.2 | 4215 | UTR 3 | XP_011509699.1 | |||
XM_011511398.2 | 4215 | Intron | XP_011509700.1 | |||
XM_011511399.2 | 4215 | UTR 3 | XP_011509701.1 | |||
XM_011511400.2 | 4215 | UTR 3 | XP_011509702.1 | |||
XM_011511401.2 | 4215 | Intron | XP_011509703.1 | |||
XM_017004426.1 | 4215 | UTR 3 | XP_016859915.1 | |||
XM_017004427.1 | 4215 | UTR 3 | XP_016859916.1 | |||
XM_017004428.1 | 4215 | UTR 3 | XP_016859917.1 |