Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INO80C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INO80C - INO80 complex subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098817.1 | 825 | UTR 3 | NP_001092287.1 | |||
NM_001308064.1 | 825 | UTR 3 | NP_001294993.1 | |||
NM_194281.3 | 825 | UTR 3 | NP_919257.2 | |||
XM_005258196.4 | 825 | UTR 3 | XP_005258253.1 | |||
XM_005258197.4 | 825 | UTR 3 | XP_005258254.1 | |||
XM_011525812.2 | 825 | UTR 3 | XP_011524114.1 | |||
XM_017025549.1 | 825 | UTR 3 | XP_016881038.1 |