Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601619 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGDS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RALGDS - ral guanine nucleotide dissociation stimulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042368.2 | 3121 | UTR 3 | NP_001035827.1 | |||
NM_001271774.1 | 3121 | UTR 3 | NP_001258703.1 | |||
NM_001271775.1 | 3121 | UTR 3 | NP_001258704.1 | |||
NM_001271776.1 | 3121 | UTR 3 | NP_001258705.1 | |||
NM_006266.3 | 3121 | UTR 3 | NP_006257.1 |
Set Membership: |
HapMap |