Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPSAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPSAP2 - phosphoribosyl pyrophosphate synthetase associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243936.1 | Intron | NP_001230865.1 | ||||
NM_001243940.1 | Intron | NP_001230869.1 | ||||
NM_001243941.1 | Intron | NP_001230870.1 | ||||
NM_001243942.1 | Intron | NP_001230871.1 | ||||
NM_002767.3 | Intron | NP_002758.1 | ||||
XM_005256725.2 | Intron | XP_005256782.1 | ||||
XM_005256726.1 | Intron | XP_005256783.1 | ||||
XM_005256729.4 | Intron | XP_005256786.1 | ||||
XM_017024864.1 | Intron | XP_016880353.1 | ||||
XM_017024865.1 | Intron | XP_016880354.1 | ||||
XM_017024866.1 | Intron | XP_016880355.1 | ||||
XM_017024867.1 | Intron | XP_016880356.1 | ||||
XM_017024868.1 | Intron | XP_016880357.1 | ||||
XM_017024869.1 | Intron | XP_016880358.1 | ||||
XM_017024870.1 | Intron | XP_016880359.1 | ||||
XM_017024871.1 | Intron | XP_016880360.1 | ||||
XM_017024872.1 | Intron | XP_016880361.1 | ||||
XM_017024873.1 | Intron | XP_016880362.1 | ||||
XM_017024874.1 | Intron | XP_016880363.1 | ||||
XM_017024875.1 | Intron | XP_016880364.1 | ||||
XM_017024876.1 | Intron | XP_016880365.1 |