Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607222 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD16 - ankyrin repeat domain 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009941.2 | 22 | Intron | NP_001009941.1 | |||
NM_001009943.2 | 22 | Intron | NP_001009943.1 | |||
NM_019046.2 | 22 | Intron | NP_061919.1 |
FBXO18 - F-box protein, helicase, 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258452.1 | 22 | Intron | NP_001245381.1 | |||
NM_001258453.1 | 22 | UTR 5 | NP_001245382.1 | |||
NM_032807.4 | 22 | Intron | NP_116196.3 | |||
NM_178150.2 | 22 | UTR 5 | NP_835363.1 | |||
XM_011519748.2 | 22 | UTR 5 | XP_011518050.1 | |||
XM_017016842.1 | 22 | Intron | XP_016872331.1 | |||
XM_017016843.1 | 22 | Intron | XP_016872332.1 | |||
XM_017016844.1 | 22 | UTR 5 | XP_016872333.1 | |||
XM_017016845.1 | 22 | Intron | XP_016872334.1 | |||
XM_017016846.1 | 22 | Intron | XP_016872335.1 |
Set Membership: |
HapMap |