Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608234 MIM: 605591 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAL3ST3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GAL3ST3 - galactose-3-O-sulfotransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SF3B2 - splicing factor 3b subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006842.2 | 441 | Intron | NP_006833.2 | |||
XM_005273726.3 | 441 | UTR 5 | XP_005273783.1 | |||
XM_011544740.2 | 441 | UTR 5 | XP_011543042.1 | |||
XM_017017144.1 | 441 | UTR 5 | XP_016872633.1 |
Set Membership: |
HapMap |