Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGAGGCAGGAACCAACGTACCA[C/T]ATTTTTTGTACAAATTGTTGGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602095 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376714 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105376714 - uncharacterized LOC105376714 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC30A4 - solute carrier family 30 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321036.1 | 2345 | Intron | NP_001307965.1 | |||
NM_013309.5 | 2345 | UTR 3 | NP_037441.2 | |||
XM_011521997.2 | 2345 | UTR 3 | XP_011520299.1 | |||
XM_017022560.1 | 2345 | UTR 3 | XP_016878049.1 | |||
XM_017022561.1 | 2345 | Intron | XP_016878050.1 | |||
XM_017022562.1 | 2345 | Intron | XP_016878051.1 |
Set Membership: |
HapMap |