Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602453 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGAD - integrin subunit alpha D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318185.1 | Intron | NP_001305114.1 | ||||
NM_005353.2 | Intron | NP_005344.2 | ||||
XM_011545835.2 | Intron | XP_011544137.1 | ||||
XM_011545836.2 | Intron | XP_011544138.1 | ||||
XM_011545837.2 | Intron | XP_011544139.1 | ||||
XM_011545838.2 | Intron | XP_011544140.1 | ||||
XM_011545839.2 | Intron | XP_011544141.1 | ||||
XM_011545841.2 | Intron | XP_011544143.1 | ||||
XM_011545842.2 | Intron | XP_011544144.1 | ||||
XM_011545843.2 | Intron | XP_011544145.1 | ||||
XM_011545844.2 | Intron | XP_011544146.1 | ||||
XM_011545845.2 | Intron | XP_011544147.1 | ||||
XM_011545846.2 | Intron | XP_011544148.1 | ||||
XM_011545847.2 | Intron | XP_011544149.1 | ||||
XM_011545848.2 | Intron | XP_011544150.1 | ||||
XM_017023215.1 | Intron | XP_016878704.1 |
Set Membership: |
HapMap |