Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616017 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM69 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIM69 - tripartite motif containing 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301144.1 | 386 | Intron | NP_001288073.1 | |||
NM_001301145.1 | 386 | Intron | NP_001288074.1 | |||
NM_001301146.1 | 386 | Intron | NP_001288075.1 | |||
NM_080745.4 | 386 | Intron | NP_542783.2 | |||
NM_182985.4 | 386 | Missense Mutation | AAC,GAC | N,D 15 | NP_892030.3 |
Set Membership: |
HapMap |