Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609212 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASRGL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASRGL1 - asparaginase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083926.1 | 19 | UTR 5 | NP_001077395.1 | |||
NM_025080.3 | 19 | UTR 5 | NP_079356.3 | |||
XM_005274305.3 | 19 | Intron | XP_005274362.1 | |||
XM_005274306.2 | 19 | Intron | XP_005274363.1 | |||
XM_011545265.2 | 19 | UTR 5 | XP_011543567.1 | |||
XM_011545266.2 | 19 | UTR 5 | XP_011543568.1 | |||
XM_017018354.1 | 19 | UTR 5 | XP_016873843.1 |
Set Membership: |
HapMap |