Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607997 MIM: 606553 | ||||||||||||||||||||
Literature Links: |
NPW PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPW - neuropeptide W | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A3R2 - SLC9A3 regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130012.2 | Intron | NP_001123484.1 | ||||
NM_001252073.1 | Intron | NP_001239002.1 | ||||
NM_001252075.1 | Intron | NP_001239004.1 | ||||
NM_001252076.1 | Intron | NP_001239005.1 | ||||
NM_004785.5 | Intron | NP_004776.3 | ||||
XM_005255702.4 | Intron | XP_005255759.1 | ||||
XM_006720981.1 | Intron | XP_006721044.1 | ||||
XM_017023894.1 | Intron | XP_016879383.1 | ||||
XM_017023895.1 | Intron | XP_016879384.1 |