Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606998 MIM: 607593 MIM: 191130 | ||||||||||||||||||||
Literature Links: |
FLOT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLOT1 - flotillin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318875.1 | 1941 | UTR 3 | NP_001305804.1 | |||
NM_005803.3 | 1941 | UTR 3 | NP_005794.1 | |||
XM_005248780.3 | 1941 | Intron | XP_005248837.1 | |||
XM_006714947.3 | 1941 | Intron | XP_006715010.1 | |||
XM_017010157.1 | 1941 | Intron | XP_016865646.1 | |||
XM_017010158.1 | 1941 | Intron | XP_016865647.1 |
MDC1 - mediator of DNA damage checkpoint 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBB - tubulin beta class I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |