Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 605701 | ||||||||||||||||||||||||||
Literature Links: |
TRIM10 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TRIM10 - tripartite motif containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006778.3 | 804 | Intron | NP_006769.2 | |||
NM_052828.2 | 804 | Intron | NP_439893.2 | |||
XM_011514221.1 | 804 | Intron | XP_011512523.1 | |||
XM_011514222.2 | 804 | Intron | XP_011512524.1 | |||
XM_011514223.2 | 804 | Intron | XP_011512525.1 | |||
XM_011514225.1 | 804 | Intron | XP_011512527.1 |
TRIM15 - tripartite motif containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033229.2 | 804 | Missense Mutation | CCC,TCC | P,S 109 | NP_150232.2 | |
XM_011514987.1 | 804 | Missense Mutation | CCC,TCC | P,S 4 | XP_011513289.1 | |
XM_011514988.2 | 804 | UTR 5 | XP_011513290.1 |
Set Membership: |
HapMap |