Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607333 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALS2CR11 - amyotrophic lateral sclerosis 2 chromosome region candidate 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168216.1 | Intron | NP_001161688.1 | ||||
NM_001168217.1 | Intron | NP_001161689.1 | ||||
NM_001168221.1 | Intron | NP_001161693.1 | ||||
NM_152525.5 | Intron | NP_689738.3 | ||||
XM_006712331.3 | Intron | XP_006712394.1 | ||||
XM_006712332.3 | Intron | XP_006712395.1 | ||||
XM_006712333.3 | Intron | XP_006712396.1 | ||||
XM_006712334.3 | Intron | XP_006712397.1 | ||||
XM_006712335.3 | Intron | XP_006712398.1 | ||||
XM_006712336.3 | Intron | XP_006712399.1 | ||||
XM_011510736.2 | Intron | XP_011509038.1 |
STRADB - STE20-related kinase adaptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |