Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608435 | ||||||||||||||||||||
Literature Links: |
MRAS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRAS - muscle RAS oncogene homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085049.2 | 653 | Intron | NP_001078518.1 | |||
NM_001252090.1 | 653 | Intron | NP_001239019.1 | |||
NM_001252091.1 | 653 | UTR 5 | NP_001239020.1 | |||
NM_001252092.1 | 653 | Intron | NP_001239021.1 | |||
NM_001252093.1 | 653 | Intron | NP_001239022.1 | |||
NM_012219.4 | 653 | UTR 5 | NP_036351.3 | |||
XM_005247228.1 | 653 | Intron | XP_005247285.1 | |||
XM_005247229.2 | 653 | Intron | XP_005247286.1 | |||
XM_017005887.1 | 653 | Intron | XP_016861376.1 |