Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCACCAGGCTCTCATGGCTCAG[C/T]CAAGGGGCAAGTCTTCTCAGCTCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138200 MIM: 609172 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPT - glutamic--pyruvic transaminase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928953 - uncharacterized LOC101928953 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R16A - protein phosphatase 1 regulatory subunit 16A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005272350.2 | Intron | XP_005272407.1 | ||||
XM_011517349.2 | Intron | XP_011515651.1 | ||||
XM_017013912.1 | Intron | XP_016869401.1 | ||||
XM_017013913.1 | Intron | XP_016869402.1 | ||||
XM_017013914.1 | Intron | XP_016869403.1 | ||||
XM_017013915.1 | Intron | XP_016869404.1 |
Set Membership: |
HapMap |