Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609803 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OSGEPL1 - O-sialoglycoprotein endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022353.2 | 2739 | UTR 3 | NP_071748.2 | |||
XM_005246766.3 | 2739 | UTR 3 | XP_005246823.1 | |||
XM_006712685.1 | 2739 | UTR 3 | XP_006712748.1 | |||
XM_006712686.1 | 2739 | UTR 3 | XP_006712749.1 | |||
XM_011511631.1 | 2739 | UTR 3 | XP_011509933.1 | |||
XM_017004676.1 | 2739 | Intron | XP_016860165.1 | |||
XM_017004677.1 | 2739 | Intron | XP_016860166.1 | |||
XM_017004678.1 | 2739 | UTR 3 | XP_016860167.1 | |||
XM_017004679.1 | 2739 | Intron | XP_016860168.1 | |||
XM_017004680.1 | 2739 | Intron | XP_016860169.1 | |||
XM_017004681.1 | 2739 | UTR 3 | XP_016860170.1 | |||
XM_017004682.1 | 2739 | UTR 3 | XP_016860171.1 | |||
XM_017004683.1 | 2739 | UTR 3 | XP_016860172.1 | |||
XM_017004684.1 | 2739 | UTR 3 | XP_016860173.1 | |||
XM_017004685.1 | 2739 | Intron | XP_016860174.1 |
Set Membership: |
HapMap |