Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNN2 - potassium calcium-activated channel subfamily N member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278204.1 | Intron | NP_001265133.1 | ||||
NM_021614.3 | Intron | NP_067627.2 | ||||
NM_170775.2 | Intron | NP_740721.1 | ||||
XM_011543387.1 | Intron | XP_011541689.1 | ||||
XM_011543389.1 | Intron | XP_011541691.1 | ||||
XM_017009457.1 | Intron | XP_016864946.1 |