Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605879 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNN2 - potassium calcium-activated channel subfamily N member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278204.1 | Intron | NP_001265133.1 | ||||
NM_021614.3 | Intron | NP_067627.2 | ||||
NM_170775.2 | Intron | NP_740721.1 | ||||
XM_011543387.1 | Intron | XP_011541689.1 | ||||
XM_011543389.1 | Intron | XP_011541691.1 | ||||
XM_017009457.1 | Intron | XP_016864946.1 |
Set Membership: |
HapMap |