Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614697 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HDHD2 - haloacid dehalogenase like hydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318765.1 | Intron | NP_001305694.1 | ||||
NM_032124.4 | Intron | NP_115500.1 | ||||
XM_005258371.4 | Intron | XP_005258428.1 | ||||
XM_006722557.2 | Intron | XP_006722620.1 | ||||
XM_011526227.2 | Intron | XP_011524529.1 | ||||
XM_011526228.2 | Intron | XP_011524530.1 | ||||
XM_017026039.1 | Intron | XP_016881528.1 | ||||
XM_017026040.1 | Intron | XP_016881529.1 |
KATNAL2 - katanin catalytic subunit A1 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |