Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147679 | ||||||||||||||||||||
Literature Links: |
IL1RN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL1RN - interleukin 1 receptor antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000577.4 | Intron | NP_000568.1 | ||||
NM_001318914.1 | Intron | NP_001305843.1 | ||||
NM_173841.2 | Intron | NP_776213.1 | ||||
NM_173842.2 | Intron | NP_776214.1 | ||||
NM_173843.2 | Intron | NP_776215.1 | ||||
XM_005263661.4 | Intron | XP_005263718.1 | ||||
XM_011511121.1 | Intron | XP_011509423.1 |