Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGACTGGCCGGGGTACAAATGGTT[C/T]TGAAGCACTTGAAGCTCACAGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130207 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100130207 - uncharacterized LOC100130207 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRN1 - leucine rich repeat neuronal 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324188.1 | 641 | UTR 5 | NP_001311117.1 | |||
NM_001324189.1 | 641 | UTR 5 | NP_001311118.1 | |||
NM_020873.6 | 641 | Intron | NP_065924.3 |